accessibility menu, dialog, popup

UserName

HOFFF

CAS RN 461-18-7

IUPAC Name: 4,4,4-trifluorobutan-1-ol
Synonyms: 4,4,4-trifluoro-1-butanol
Molecular Weight (g/mol): 128.09
Molecular Formula: C4H7F3O
InChi Key: VKRFUGHXKNNIJO-UHFFFAOYSA-N
SMILES: OCCCC(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)