missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(4-methoxyphenyl)-1,3-butanedione 98.0+%, TCI America™

Catalog No. p-7137675
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32831G 1 g
T32835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32831G Supplier TCI America Supplier No. T32831G

Chemical Identifiers

CAS 15191-68-1
Molecular Formula C11H9F3O3
Molecular Weight (g/mol) 246.185
MDL Number MFCD03238723
InChI Key NNEIYSHJFCLFES-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-4-methoxyphenyl butane-1,3-dione, 4,4,4-trifluoro-1-4-methoxyphenyl-1,3-butanedione, 2,4-butanedione, 1,1,1-trifluoro-4-4-methoxyphenyl, 4-methoxybenzoyl-1,1,1-trifluoroacetone, 1,3-butanedione,4,4,4-trifluoro-1-4-methoxyphenyl, tos-bb-0662, p-methoxybenzoyltrifluoroacetone, 1-p-methoxybenzoyl-3,3,3-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-4-methoxyphenyl, 1-4-methoxy-phenyl-4,4,4-trifluoro-butane-1,3-dione
PubChem CID 139903
IUPAC Name 4,4,4-trifluoro-1-(4-methoxyphenyl)butane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 1 g
Formula Weight 246.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,4-Trifluoro-1-(4-methoxyphenyl)-1,3-butanedione