accessibility menu, dialog, popup

UserName

CAS RN 15191-68-1

IUPAC Name: 4,4,4-trifluoro-1-(4-methoxyphenyl)butane-1,3-dione
Synonyms: 4,4,4-trifluoro-1-(4-methoxyphenyl)butane-1,3-dione
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: NNEIYSHJFCLFES-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results