missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione, 99%

Catalog No. p-7024307
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1872606 5 g
AAA1872614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1872606 Supplier Thermo Scientific Chemicals Supplier No. A1872606

CAS: 893-33-4 | C14H9F3O2 | 266.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 893-33-4
Molecular Formula C14H9F3O2
Molecular Weight (g/mol) 266.219
MDL Number MFCD00054501
InChI Key WVVLURYIQCXPIV-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione, 1-2-naphthoyl-3,3,3-trifluoroacetone, 4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione, 3-2-naphthoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl, 2-naphthoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl, 4,4,4-trifluoro-3-oxo-2'-butyronaphthone, acmc-1bl5t
PubChem CID 70179
IUPAC Name 4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F

Specifications

Melting Point 74°C to 76°C
Quantity 5 g
Formula Weight 266.22
Percent Purity 99%
Chemical Name or Material 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only