missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid, 97%

Catalog No. p-7036097
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54342MD 250 mg
AAH5434203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54342MD Supplier Thermo Scientific Chemicals Supplier No. H54342MD

CAS: 916204-05-2 | C24H23ClN2O2 | 406.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 916204-05-2
Molecular Formula C24H23ClN2O2
Molecular Weight (g/mol) 406.91
MDL Number MFCD16251290
InChI Key FSHGEKXJPYDBSO-UHFFFAOYSA-N
Synonym 4-4-4'-chlorobiphenyl-2-ylmethyl piperazin-1-yl benzoic acid, 4-4-4'-chloro-1,1'-biphenyl-2-yl methyl piperazin-1-yl benzoic acid, 4-4-4'-chlorobiphenyl-2-yl methyl piperazin-1-yl benzoic acid, 4-4-4'-chloro-2-biphenylylmethyl-1-piperazinyl benzoic acid, 4-4-4'-chloro-1,1'-biphenyl-2-yl methyl-1-piperazinyl benzoic acid, 4-4-4'-chloro 1,1'-biphenyl-2-yl methyl-1-piperazinyl benzoic acid, 4-4-4'-chloro 1,1'-biphenyl-2-yl methyl piperazin-1-yl benzoic acid, 4-4-4-chloro-1,1-biphenyl-2-yl methyl piperazin-1-yl benzoic acid
PubChem CID 53434985
IUPAC Name 4-[4-[[2-(4-chlorophenyl)phenyl]methyl]piperazin-1-yl]benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)N1CCN(CC2=CC=CC=C2C2=CC=C(Cl)C=C2)CC1

Specifications

Quantity 250 mg
Formula Weight 406.91
Percent Purity 97%
Chemical Name or Material 4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only