missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline 98.0+%, TCI America™

Catalog No. I07395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07395G 5 g
I07391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07395G Supplier TCI America Supplier No. I07395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 685103-98-4
Molecular Formula C15H18BNO2
Molecular Weight (g/mol) 255.12
MDL Number MFCD05663891
InChI Key LOMKRPABAXIQJL-UHFFFAOYSA-N
Synonym 2-(4-Isoquinolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Isoquinoline-4-boronic Acid Pinacol Ester
PubChem CID 2760594
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
SMILES CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CN=C1

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 255.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline