accessibility menu, dialog, popup

UserName

CAS RN 685103-98-4

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
Molecular Weight (g/mol): 255.12
Molecular Formula: C15H18BNO2
InChi Key: LOMKRPABAXIQJL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results