missing translation for 'onlineSavingsMsg'
Learn More

4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol 98.0+%, TCI America™

Catalog No. p-7137537
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30311G 1 g
T30315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30311G Supplier TCI America Supplier No. T30311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 760989-91-1
Molecular Formula C18H21BO3
Molecular Weight (g/mol) 296.173
MDL Number MFCD22414472
InChI Key BFQSVDDJLRMXOO-UHFFFAOYSA-N
PubChem CID 69201437
IUPAC Name 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=C(C=C3)O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
Synonym 2-(4′C-Hydroxybiphenyl-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4′C-Hydroxybiphenyl-4-yl)boronic Acid Pinacol Ester
Formula Weight 296.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol