missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide 98.0+%, TCI America™
$59.65 - $165.90
Chemical Identifiers
| CAS | 138500-85-3 |
|---|---|
| Molecular Formula | C13H18BBrO2 |
| Molecular Weight (g/mol) | 297.00 |
| MDL Number | MFCD02179493 |
| InChI Key | CBUOGMOTDGNEAW-UHFFFAOYSA-N |
| Synonym | 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl benzeneboronic acid pinacol ester, 4-bromomethylphenylboronic acid pinacol ester, 4-bromomethylphenylboronic acid, pinacol ester, 4-bromomethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaboronate, 4-bromomethyl benzeneboronicacidpinacolester, 4-bromomethyl benzeneboronic acid, pinacol ester |
| PubChem CID | 3734506 |
| IUPAC Name | 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1 |
Chemical Identifiers
| 138500-85-3 | |
| 297.00 | |
| CBUOGMOTDGNEAW-UHFFFAOYSA-N | |
| 3734506 | |
| CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1 |
| C13H18BBrO2 | |
| MFCD02179493 | |
| 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl benzeneboronic acid pinacol ester, 4-bromomethylphenylboronic acid pinacol ester, 4-bromomethylphenylboronic acid, pinacol ester, 4-bromomethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaboronate, 4-bromomethyl benzeneboronicacidpinacolester, 4-bromomethyl benzeneboronic acid, pinacol ester | |
| 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 138500-85-3 | |
| White | |
| MFCD02179493 | |
| 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl benzeneboronic acid pinacol ester, 4-bromomethylphenylboronic acid pinacol ester, 4-bromomethylphenylboronic acid, pinacol ester, 4-bromomethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaboronate, 4-bromomethyl benzeneboronicacidpinacolester, 4-bromomethyl benzeneboronic acid, pinacol ester | |
| CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1 | |
| 297.00 | |
| 297.00 | |
| Crystalline Powder |
| 86°C | |
| C13H18BBrO2 | |
| 1 g | |
| CBUOGMOTDGNEAW-UHFFFAOYSA-N | |
| 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 3734506 | |
| ≥98.0% (GC,T) | |
| 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide |
Safety and Handling
TSCA : No