missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide 98.0+%, TCI America™

Catalog No. T29451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29451G 1 g
T29455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29451G Supplier TCI America Supplier No. T29451G

Chemical Identifiers

CAS 138500-85-3
Molecular Formula C13H18BBrO2
Molecular Weight (g/mol) 297.00
MDL Number MFCD02179493
InChI Key CBUOGMOTDGNEAW-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl benzeneboronic acid pinacol ester, 4-bromomethylphenylboronic acid pinacol ester, 4-bromomethylphenylboronic acid, pinacol ester, 4-bromomethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaboronate, 4-bromomethyl benzeneboronicacidpinacolester, 4-bromomethyl benzeneboronic acid, pinacol ester
PubChem CID 3734506
IUPAC Name 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1

Specifications

Melting Point 86°C
Color White
Quantity 1 g
Formula Weight 297.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide