missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7137771
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
T34961G 1 g
T34965G 5 g
2 options

Catalog No. T34961G

Supplier: TCI America T34961G

Only null left

Chemical Identifiers

CAS 302348-51-2
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.102
MDL Number MFCD09837617
InChI Key GZZBZWITJNATOD-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethyl phenylboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 4-hydroxymethylphenylboronic acid pinacol ester, 4-hydroxymethyl benzeneboronic acid pinacol ester, 2-4-hydroxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzenemethanol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl alcohol, amtb135
PubChem CID 11402050
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CO

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Alcohol
TSCA No
Recommended Storage Refrigerator