missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 97.0+%, TCI America™

Catalog No. T29231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29231G 1 g
T29235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29231G Supplier TCI America Supplier No. T29231G

Chemical Identifiers

CAS 128376-64-7
Molecular Formula C13H17BO3
Molecular Weight (g/mol) 232.09
MDL Number MFCD04972375
InChI Key DMBMXJJGPXADPO-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester
PubChem CID 2769536
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 1 g
Formula Weight 232.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde