missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole 98.0+%, TCI America™
$98.79 - $300.11
Chemical Identifiers
| CAS | 171364-79-7 |
|---|---|
| Molecular Formula | C13H19BO3 |
| Molecular Weight (g/mol) | 234.10 |
| MDL Number | MFCD05663864 |
| InChI Key | VFIKPDSQDNROGM-UHFFFAOYSA-N |
| Synonym | 4-Methoxyphenylboronic Acid Pinacol Ester, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| PubChem CID | 584319 |
| IUPAC Name | 2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
Chemical Identifiers
| 171364-79-7 | |
| 234.10 | |
| VFIKPDSQDNROGM-UHFFFAOYSA-N | |
| 584319 | |
| COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
| C13H19BO3 | |
| MFCD05663864 | |
| 4-Methoxyphenylboronic Acid Pinacol Ester, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 171364-79-7 | |
| C13H19BO3 | |
| 5 g | |
| VFIKPDSQDNROGM-UHFFFAOYSA-N | |
| 2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 584319 | |
| ≥98.0% (GC,T) | |
| 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole |
| Yellow | |
| MFCD05663864 | |
| 4-Methoxyphenylboronic Acid Pinacol Ester, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
| 234.10 | |
| 234.10 | |
| Liquid |
Safety and Handling
TSCA : No