CAS RN 171364-79-7
IUPAC Name:
2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
234.1
Molecular Formula:
C13H19BO3
InChi Key:
VFIKPDSQDNROGM-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole 98.0+%, TCI America™
CAS: 171364-79-7 Molecular Formula: C13H19BO3 Molecular Weight (g/mol): 234.10 MDL Number: MFCD05663864 InChI Key: VFIKPDSQDNROGM-UHFFFAOYSA-N Synonym: 4-Methoxyphenylboronic Acid Pinacol Ester, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane PubChem CID: 584319 IUPAC Name: 2-(4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: COC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1