missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 234.10 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole |
Safety and Handling
| TSCA | No |