missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 98.0+%, TCI America™
Supplier: TCI America T3400200MG
Specifications
| 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine | |
| 112°C | |
| C17H20BNO2 | |
| 200 mg | |
| PTNMCYWJKRZCDE-UHFFFAOYSA-N | |
| 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine | |
| 58526497 | |
| ≥98.0% (GC,T) |
| 1009033-87-7 | |
| White-Yellow | |
| MFCD12828171 | |
| 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 4-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=NC=C3 | |
| 281.162 | |
| 281.16 | |
| Crystalline Powder |
Chemical Identifiers
| 1009033-87-7 | |
| 281.162 | |
| PTNMCYWJKRZCDE-UHFFFAOYSA-N | |
| 58526497 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=NC=C3 |
| C17H20BNO2 | |
| MFCD12828171 | |
| 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 4-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine |
Safety and Handling
TSCA : No