missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 98.0+%, TCI America™
$195.73 - $579.71
Chemical Identifiers
| CAS | 1009033-87-7 |
|---|---|
| Molecular Formula | C17H20BNO2 |
| Molecular Weight (g/mol) | 281.162 |
| MDL Number | MFCD12828171 |
| InChI Key | PTNMCYWJKRZCDE-UHFFFAOYSA-N |
| Synonym | 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 4-(4-Pyridyl)phenylboronic Acid Pinacol Ester |
| PubChem CID | 58526497 |
| IUPAC Name | 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=NC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3400200MG
|
TCI America
T3400200MG |
200 mg |
Each for $195.73
|
|
|||||
|
T34001G
|
TCI America
T34001G |
1 g |
Each for $579.71
|
|
|||||
Chemical Identifiers
| 1009033-87-7 | |
| 281.162 | |
| PTNMCYWJKRZCDE-UHFFFAOYSA-N | |
| 58526497 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=NC=C3 |
| C17H20BNO2 | |
| MFCD12828171 | |
| 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 4-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine |
Specifications
| 1009033-87-7 | |
| White-Yellow | |
| MFCD12828171 | |
| 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 4-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=NC=C3 | |
| 281.162 | |
| 281.16 | |
| Crystalline Powder |
| 112°C | |
| C17H20BNO2 | |
| 200 mg | |
| PTNMCYWJKRZCDE-UHFFFAOYSA-N | |
| 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine | |
| 58526497 | |
| ≥98.0% (GC,T) | |
| 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine |
Safety and Handling
TSCA : No