missing translation for 'onlineSavingsMsg'
Learn More

4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%

Catalog No. AC446540010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446540010 1 g
AC446540050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446540010 Supplier Thermo Scientific Chemicals Supplier No. 446540010
Only null left

CAS: 16063-80-2 | C5H5F6NO2 | 225.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16063-80-2
Molecular Formula C5H5F6NO2
Molecular Weight (g/mol) 225.09
MDL Number MFCD00014349
InChI Key KRNSHCKTGFAMPQ-UHFFFAOYSA-N
IUPAC Name 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
SMILES NC(C(C(F)(F)F)C(F)(F)F)C(O)=O

Specifications

Melting Point 207°C to 211°C
Assay Percent Range 96 to 104%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in mineral acids
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 4,4,4,4',4',4'-Hexafluoro-DL-valine
Hazard Category GHS Signal Word: Warning
EINECSNumber 240-207-5

RUO – Research Use Only