accessibility menu, dialog, popup

UserName

CAS RN 16063-80-2

NH2FFFFFFOHO

CAS RN 16063-80-2

IUPAC Name: 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Synonyms: 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Molecular Weight (g/mol): 225.09
Molecular Formula: C5H5F6NO2
InChi Key: KRNSHCKTGFAMPQ-UHFFFAOYSA-N
SMILES: NC(C(C(F)(F)F)C(F)(F)F)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)