missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-(1,3-Dimethylbutylidene)diphenol 98.0+%, TCI America™
$93.21 - $93.21
Chemical Identifiers
| CAS | 6807-17-6 |
|---|---|
| Molecular Formula | C18H22O2 |
| Molecular Weight (g/mol) | 270.372 |
| InChI Key | VHLLJTHDWPAQEM-UHFFFAOYSA-N |
| Synonym | 2,2-Bis(4-hydroxyphenyl)-4-methylpentane |
| PubChem CID | 81259 |
| IUPAC Name | 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol |
| SMILES | CC(C)CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O |
Chemical Identifiers
| 6807-17-6 | |
| 270.372 | |
| 2,2-Bis(4-hydroxyphenyl)-4-methylpentane | |
| 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol |
| C18H22O2 | |
| VHLLJTHDWPAQEM-UHFFFAOYSA-N | |
| 81259 | |
| CC(C)CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O |
Specifications
| 6807-17-6 | |
| White-Yellow | |
| 25 g | |
| 2,2-Bis(4-hydroxyphenyl)-4-methylpentane | |
| CC(C)CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O | |
| 270.372 | |
| 270.37 | |
| Crystalline Powder |
| 154°C | |
| C18H22O2 | |
| 3077 | |
| VHLLJTHDWPAQEM-UHFFFAOYSA-N | |
| 4-[2-(4-hydroxyphenyl)-4-methylpentan-2-yl]phenol | |
| 81259 | |
| ≥98.0% (GC) | |
| 4,4′-(1,3-Dimethylbutylidene)diphenol |
Safety and Handling
EINECSNumber : (4)-1709
TSCA : Yes