missing translation for 'onlineSavingsMsg'
Learn More

4(3H)-Pyrimidinone 98.0+%, TCI America™

Catalog No. H10001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10001G 1 g
H10005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10001G Supplier TCI America Supplier No. H10001G

Chemical Identifiers

CAS 4562-27-0
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00167178
InChI Key DNCYBUMDUBHIJZ-UHFFFAOYSA-N
Synonym pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
PubChem CID 20695
IUPAC Name 1H-pyrimidin-6-one
SMILES C1=CN=CNC1=O

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 1 g
Formula Weight 96.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4(3H)-Pyrimidinone
RTECSNumber UW7350100
TSCA Yes