missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine Hydrochloride 98.0+%, TCI America™

Catalog No. T3448200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3448200MG 200 mg
T34481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3448200MG Supplier TCI America Supplier No. T3448200MG

Chemical Identifiers

CAS 1258874-29-1
Molecular Formula C9H9ClF3N3
Molecular Weight (g/mol) 251.637
InChI Key DNTDGTWJZOMTAK-UHFFFAOYSA-N
Synonym 3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine Hydrochloride
PubChem CID 66870186
IUPAC Name [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)C2(N=N2)C(F)(F)F.Cl

Specifications

Melting Point 142°C
Color White
Quantity 200 mg
Formula Weight 251.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine Hydrochloride
TSCA No
Recommended Storage Refrigerator