CAS RN 1258874-29-1
CAS RN 1258874-29-1
IUPAC Name:
1-{4-[3-(trifluoromethyl)-3H-diazirin-3-yl]phenyl}methanamine hydrochloride
Synonyms:
1-{4-[3-(trifluoromethyl)diazirin-3-yl]phenyl}methanamine hydrochloride
Molecular Weight (g/mol):
251.64
Molecular Formula:
C9H9ClF3N3
InChi Key:
DNTDGTWJZOMTAK-UHFFFAOYSA-N
SMILES:
Cl.NCC1=CC=C(C=C1)C1(N=N1)C(F)(F)F
1
–
1
of
1
results
1
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine Hydrochloride 98.0+%, TCI America™
CAS: 1258874-29-1 Molecular Formula: C9H9ClF3N3 Molecular Weight (g/mol): 251.637 InChI Key: DNTDGTWJZOMTAK-UHFFFAOYSA-N Synonym: 3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine Hydrochloride PubChem CID: 66870186 IUPAC Name: [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanamine;hydrochloride SMILES: C1=CC(=CC=C1CN)C2(N=N2)C(F)(F)F.Cl