missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid 98.0+%, TCI America™

Supplier:  TCI America T28201G

 View more versions of this product

Catalog No. T28201G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid
125°C
C9H5F3N2O2
1 g
C1=CC(=CC=C1C(=O)O)C2(N=N2)C(F)(F)F
230.146
230.15
Crystalline Powder
85559-46-2
White
MFCD00236809
CZPAJVBVULSLGG-UHFFFAOYSA-N
4-[3-(trifluoromethyl)diazirin-3-yl]benzoic acid
128718
≥98.0% (HPLC,T)

Chemical Identifiers

85559-46-2
230.146
CZPAJVBVULSLGG-UHFFFAOYSA-N
4-[3-(trifluoromethyl)diazirin-3-yl]benzoic acid
C9H5F3N2O2
MFCD00236809
128718
C1=CC(=CC=C1C(=O)O)C2(N=N2)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions