missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid 98.0+%, TCI America™

Catalog No. p-7137436
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2820200MG 200 mg
T28201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2820200MG Supplier TCI America Supplier No. T2820200MG
Only null left

Chemical Identifiers

CAS 85559-46-2
Molecular Formula C9H5F3N2O2
Molecular Weight (g/mol) 230.146
MDL Number MFCD00236809
InChI Key CZPAJVBVULSLGG-UHFFFAOYSA-N
PubChem CID 128718
IUPAC Name 4-[3-(trifluoromethyl)diazirin-3-yl]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C2(N=N2)C(F)(F)F

Specifications

Melting Point 125°C
Color White
Quantity 200 mg
Formula Weight 230.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid
TSCA No
Recommended Storage Refrigerator