missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Pyrrolidinyl)morpholine, 97%

Catalog No. AAH5100203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5100203 1 g
AAH51002MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5100203 Supplier Thermo Scientific Chemicals Supplier No. H5100203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53617-37-1 | C8H16N2O | 156.23 g/mol

4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 53617-37-1
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.23
MDL Number MFCD06740352
InChI Key OPJDRNMJJNFSNZ-UHFFFAOYNA-N
Synonym 4-pyrrolidin-3-yl morpholine, 4-pyrrolidin-3-yl-morpholine, 4-3-pyrrolidinyl morpholine, 4-3-pyrrolidino morpholine, acmc-20aiao, pubchem11379, 3-morpholin-4-yl pyrrolidine, morpholine, 4-3-pyrrolidinyl
PubChem CID 14627373
IUPAC Name 4-pyrrolidin-3-ylmorpholine
SMILES C1CC(CN1)N1CCOCC1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 4-(3-Pyrrolidinyl)morpholine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only