CAS RN 53617-37-1
IUPAC Name:
4-(pyrrolidin-3-yl)morpholine
Synonyms:
4-(pyrrolidin-3-yl)morpholine
Molecular Weight (g/mol):
156.23
Molecular Formula:
C8H16N2O
InChi Key:
OPJDRNMJJNFSNZ-UHFFFAOYNA-N
SMILES:
C1CC(CN1)N1CCOCC1
1
–
1
of
1
results
1
Promotions Available
4-(3-Pyrrolidinyl)morpholine, 97%
4-(3-Pyrrolidinyl)morpholine, 97%
CAS: 53617-37-1 Molecular Formula: C8H16N2O Molecular Weight (g/mol): 156.23 MDL Number: MFCD06740352 InChI Key: OPJDRNMJJNFSNZ-UHFFFAOYNA-N Synonym: 4-pyrrolidin-3-yl morpholine,4-pyrrolidin-3-yl-morpholine,4-3-pyrrolidinyl morpholine,4-3-pyrrolidino morpholine,acmc-20aiao,pubchem11379,3-morpholin-4-yl pyrrolidine,morpholine, 4-3-pyrrolidinyl PubChem CID: 14627373 IUPAC Name: 4-pyrrolidin-3-ylmorpholine SMILES: C1CC(CN1)N1CCOCC1