missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine, 97%

Catalog No. p-7033716
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50873MD 250 mg
AAH5087303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50873MD Supplier Thermo Scientific Chemicals Supplier No. H50873MD
Only null left

CAS: 733748-92-0 | C10H17N3O | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 733748-92-0
Molecular Formula C10H17N3O
Molecular Weight (g/mol) 195.27
MDL Number MFCD08061038
InChI Key QDRJZNBKSMAEIE-UHFFFAOYSA-N
Synonym 4-3-isopropyl-1,2,4-oxadiazol-5-yl piperidine, 3-isopropyl-5-piperidin-4-yl-1,2,4-oxadiazole, 4-3-isopropyl-1,2,4 oxadiazol-5-yl-piperidine, 5-piperidin-4-yl-3-propan-2-yl-1,2,4-oxadiazole, piperidine, 4-3-1-methylethyl-1,2,4-oxadiazol-5-yl, 4-3-propan-2-yl-1,2,4-oxadiazol-5-yl piperidine, 4-3-1-methylethyl-1,2,4-oxadiazol-5-yl piperidine, 3-isopropyl-5-4-piperidyl-1,2,4-oxadiazole, 3-methylethyl-5-4-piperidyl-1,2,4-oxadiazole
PubChem CID 24208845
IUPAC Name 5-piperidin-4-yl-3-propan-2-yl-1,2,4-oxadiazole
SMILES CC(C)C1=NOC(=N1)C1CCNCC1

Specifications

Refractive Index 1.486
Quantity 250 mg
Formula Weight 195.27
Percent Purity 97%
Chemical Name or Material 4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only