missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(3-Hydroxyphenyl)piperidine 98.0+%, TCI America™
Supplier: TCI America H096525G
Specifications
4-(3-Hydroxyphenyl)piperidine | |
225°C | |
C11H15NO | |
25 g | |
FINLIMGQGJZNRN-UHFFFAOYSA-N | |
3-piperidin-4-ylphenol | |
10583453 | |
≥98.0% (GC) |
110878-71-2 | |
White-Yellow | |
MFCD02093474 | |
3-(4-Piperidyl)phenol | |
C1CNCCC1C2=CC(=CC=C2)O | |
177.247 | |
177.25 | |
Crystalline Powder |
Chemical Identifiers
110878-71-2 | |
177.247 | |
FINLIMGQGJZNRN-UHFFFAOYSA-N | |
10583453 | |
C1CNCCC1C2=CC(=CC=C2)O |
C11H15NO | |
MFCD02093474 | |
3-(4-Piperidyl)phenol | |
3-piperidin-4-ylphenol |
Safety and Handling
TSCA : Yes