missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Hydroxyphenyl)piperidine 98.0+%, TCI America™
$1025.24 - $4061.78
Chemical Identifiers
| CAS | 110878-71-2 |
|---|---|
| Molecular Formula | C11H15NO |
| Molecular Weight (g/mol) | 177.247 |
| MDL Number | MFCD02093474 |
| InChI Key | FINLIMGQGJZNRN-UHFFFAOYSA-N |
| Synonym | 3-(4-Piperidyl)phenol |
| PubChem CID | 10583453 |
| IUPAC Name | 3-piperidin-4-ylphenol |
| SMILES | C1CNCCC1C2=CC(=CC=C2)O |
Chemical Identifiers
| 110878-71-2 | |
| 177.247 | |
| FINLIMGQGJZNRN-UHFFFAOYSA-N | |
| 10583453 | |
| C1CNCCC1C2=CC(=CC=C2)O |
| C11H15NO | |
| MFCD02093474 | |
| 3-(4-Piperidyl)phenol | |
| 3-piperidin-4-ylphenol |
Specifications
| 110878-71-2 | |
| White-Yellow | |
| MFCD02093474 | |
| 3-(4-Piperidyl)phenol | |
| C1CNCCC1C2=CC(=CC=C2)O | |
| 177.247 | |
| 177.25 | |
| Crystalline Powder |
| 225°C | |
| C11H15NO | |
| 5 g | |
| FINLIMGQGJZNRN-UHFFFAOYSA-N | |
| 3-piperidin-4-ylphenol | |
| 10583453 | |
| ≥98.0% (GC) | |
| 4-(3-Hydroxyphenyl)piperidine |
Safety and Handling
TSCA : Yes