missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chloropropyl)morpholine 98.0+%, TCI America™

Catalog No. C26025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26025G 5 g
C260225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26025G Supplier TCI America Supplier No. C26025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7357-67-7
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.645
MDL Number MFCD00039714
InChI Key PIAZYBLGBSMNLX-UHFFFAOYSA-N
Synonym 4-3-chloropropyl morpholine, n-3-chloropropyl morpholine, morpholine, 4-3-chloropropyl, 3-morpholinopropyl chloride, 1-chloro-3-morpholinopropane, 4-3-chloropropyl-morpholine, 1-chloro-3-morpholin-4-yl propane, 4-3-chloro-propyl-morpholine, morpholine, 4-3-chloropropyl-, hydrochloride
PubChem CID 95834
IUPAC Name 4-(3-chloropropyl)morpholine
SMILES C1COCCN1CCCCl

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 163.65
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 4-(3-Chloropropyl)morpholine
RTECSNumber QE0488000
TSCA No
Recommended Storage Refrigerator