accessibility menu, dialog, popup

UserName

ClNO

CAS RN 7357-67-7

IUPAC Name: 4-(3-chloropropyl)morpholine
Synonyms: 4-(3-chloropropyl)morpholine
Molecular Weight (g/mol): 163.65
Molecular Formula: C7H14ClNO
InChi Key: PIAZYBLGBSMNLX-UHFFFAOYSA-N
SMILES: ClCCCN1CCOCC1
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results