missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chloropropyl)morpholine, 95%

Catalog No. AAH3522209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3522209 10 g
AAH3522218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3522209 Supplier Thermo Scientific Chemicals Supplier No. H3522209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7357-67-7 | C7H14ClNO | 163.645 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7357-67-7
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.645
MDL Number MFCD00039714
InChI Key PIAZYBLGBSMNLX-UHFFFAOYSA-N
Synonym 4-3-chloropropyl morpholine, n-3-chloropropyl morpholine, morpholine, 4-3-chloropropyl, 3-morpholinopropyl chloride, 1-chloro-3-morpholinopropane, 4-3-chloropropyl-morpholine, 1-chloro-3-morpholin-4-yl propane, 4-3-chloro-propyl-morpholine, morpholine, 4-3-chloropropyl-, hydrochloride
PubChem CID 95834
IUPAC Name 4-(3-chloropropyl)morpholine
SMILES C1COCCN1CCCCl

Specifications

Quantity 10 g
Formula Weight 163.65
Percent Purity 95%
Chemical Name or Material 4-(3-Chloropropyl)morpholine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 461-510-0
RTECSNumber QE0488000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only