missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chlorophenoxy)piperidine, 98%, Thermo Scientific Chemicals

Catalog No. AAH5095603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5095603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5095603 Supplier Thermo Scientific Chemicals Supplier No. H5095603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 97840-40-9 | Molecular Formula: C11H14ClNO | Molecular Weight: 211.689 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97840-40-9
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061087
InChI Key BYXWGMQMUJCYPK-UHFFFAOYSA-N
Synonym 4-3-chlorophenoxy piperidine, 4-3-chloro-phenoxy-piperidine, piperidine, 4-3-chlorophenoxy, acmc-20amww, 4-m-chlorophenoxypiperidine, 4-3-chlorophenoxy-piperidine, piperidine,4-3-chlorophenoxy, 1-chloro-3-4-piperidyloxy benzene, piperidine, 4-3-chlorophenoxy-, hydrochloride
PubChem CID 2735317
IUPAC Name 4-(3-chlorophenoxy)piperidine
SMILES C1CNCCC1OC2=CC(=CC=C2)Cl

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material 4-(3-Chlorophenoxy)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only