missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one, ≥90%, Thermo Scientific™

Catalog No. CD02877CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD02877CB 250mg
CD02877DA 1g
CD02877DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD02877CB Supplier Thermo Scientific Chemicals Supplier No. CD02877CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7032-17-9
Molecular Formula C6H11N3O
Molecular Weight (g/mol) 141.174
MDL Number MFCD00053049
InChI Key LMLXMJPJCSUFAB-UHFFFAOYSA-N
Synonym 4-3-aminopropyl-2,4-dihydro-3h-pyrazol-3-one, 4-3-aminopropyl-1h-pyrazol-5 4h-one, 4-3-aminopropyl-2,4-dihydropyrazol-3-one, 4-3-aminopropyl-4,5-dihydro-1h-pyrazol-5-one, 3h-pyrazol-3-one,4-3-aminopropyl-2,4-dihydro, aminopropyldihydropyrazolone, 4-3-aminopropyl-2-pyrazolin-5-one, 4-3-aminopropyl-1,4-dihydropyrazol-5-one
PubChem CID 2776763
IUPAC Name 4-(3-aminopropyl)-1,4-dihydropyrazol-5-one
SMILES C1=NNC(=O)C1CCCN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 141.17
Percent Purity 90%
Chemical Name or Material 4-(3-Aminopropyl)-2, 4-dihydro-3H-pyrazol-3-one
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.