accessibility menu, dialog, popup

UserName

H3NNHHNO

CAS RN 7032-17-9

IUPAC Name: 3-(3-oxo-2,3-dihydro-1H-pyrazol-4-yl)propan-1-aminium
Synonyms: 3-(3-oxo-1,2-dihydropyrazol-4-yl)propan-1-aminium
Molecular Weight (g/mol): 142.18
Molecular Formula: C6H12N3O
InChi Key: JNPRSNHFVOQXKV-UHFFFAOYSA-O
SMILES: [NH3+]CCCC1=CNNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)