missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 1173020-38-6 |
| Flash Point | -17°C - Information refers to the main°Component. |
| Linear Formula | 13C6C9H24 O |
| MDL Number | MFCD09842832 |
| Synonym | 3,6,3-Nonylphenol-13C6; 363-NP-13C6; 4-(1-Ethyl-1,4-dimethylpentyl)phenol-13C6 (ring-13C6) |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 226.31 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 10 μg/mL in acetone |