missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 diethoxylate Solution, 1 μg/mL in Acetone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 1173021-42-5 |
| Flash Point | -17°C - Information refers to the main°Component. |
| Linear Formula | C19D2H30O3 |
| MDL Number | MFCD09953758 |
| Synonym | 2-{2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy-3,5-d2 ]ethoxy}ethanol; 2-{2-[4-(3,6-Dimethyl-3-heptyl)phenoxy-3,5-d2 ]ethoxy}ethanol; 3,6,3-Nonylphenol diethoxylate-d2 (ring-3,5-d2); 363-NP2 EO-D2 |
| Type | Analytical Standard |
| Quantity | 10 mL |
| Formula Weight | 310.47 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1 μg/mL in acetone |