missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33839DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC33839DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

852227-95-3
289.182
4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol
4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
C16H24BNO3
NCJDKFFODGZRRL-UHFFFAOYSA-N
4192663
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCOCC3

Specifications

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
90%
C16H24BNO3
4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCOCC3
289.182
289.18
852227-95-3
Amber Glass Bottle
5g
NCJDKFFODGZRRL-UHFFFAOYSA-N
4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
4192663
90%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents