missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 90% |
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 289.18 |
| Percent Purity | 90% |
| Chemical Name or Material | 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine |