missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 36°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 250.43 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone |
Safety and Handling
| EINECSNumber | (4)-1905 |
| TSCA | No |