accessibility menu, dialog, popup

UserName

CH3O

CAS RN 117923-32-7

IUPAC Name: 4-[2-(4-propylcyclohexyl)ethyl]cyclohexan-1-one
Synonyms: 4-[2-(4-propylcyclohexyl)ethyl]cyclohexan-1-one
Molecular Weight (g/mol): 250.43
Molecular Formula: C17H30O
InChi Key: OLRRTAGESHHXPY-UHFFFAOYSA-N
SMILES: CCCC1CCC(CCC2CCC(=O)CC2)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results