missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)butyric acid, 98%

Catalog No. p-7044639
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0756303 1 g
AAL0756306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0756303 Supplier Thermo Scientific Chemicals Supplier No. L0756303
Only null left

CAS: 4653-11-6 | C8H9O2S | 169.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4653-11-6
MDL Number MFCD00005463
ChEBI CHEBI:40114

Specifications

Density 1.169
Boiling Point 122°C (0.3 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C8H9O2S
Refractive Index 1.532
Quantity 1 g
Beilstein 127982
Synonym 4-2-thienyl butyric acid, 4-thiophen-2-yl butanoic acid, 2-thiophenebutanoic acid, 2-thiophenebutyric acid, 4-2-thienyl butanoic acid, 4-thiophen-2-yl-butyric acid, 4-thiophen-2-yl-butyricacid, gamma-alpha-thienyl butyricacid, .gamma.-.alpha.-thienyl butyric acid, 4tb
InChI Key VYTXLSQVYGNWLV-UHFFFAOYSA-M
SMILES C1=CSC(=C1)CCCC(=O)O
IUPAC Name 4-thiophen-2-ylbutanoic acid
Molecular Weight (g/mol) 169.22
PubChem CID 78386
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material 4-(2-Thienyl)butyric acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 225-090-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only