accessibility menu, dialog, popup

UserName

CAS RN 4653-11-6

IUPAC Name: 4-(thiophen-2-yl)butanoate
Synonyms: 4-(thiophen-2-yl)butanoate
Molecular Weight (g/mol): 169.22
Molecular Formula: C8H9O2S
InChi Key: VYTXLSQVYGNWLV-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCC1=CC=CS1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results