missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzenesulfonyl chloride, 96%

Catalog No. p-7032786
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34399MD 250 mg
AAH3439903 1 g
AAH3439906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH34399MD Supplier Thermo Scientific Chemicals Supplier No. H34399MD
Only null left

CAS: 209592-91-6 | C10H7ClO2S2 | 258.734 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209592-91-6
Molecular Formula C10H7ClO2S2
Molecular Weight (g/mol) 258.734
MDL Number MFCD11655569
InChI Key IRDZCMUQNGEQEP-UHFFFAOYSA-N
Synonym 4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride
PubChem CID 18387759
IUPAC Name 4-thiophen-2-ylbenzenesulfonyl chloride
SMILES C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 107°C to 108°C
Quantity 250 mg
UN Number UN3261
Formula Weight 258.75
Percent Purity 96%
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only