accessibility menu, dialog, popup

UserName

CAS RN 209592-91-6

IUPAC Name: 4-(thiophen-2-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(thiophen-2-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 258.73
Molecular Formula: C10H7ClO2S2
InChi Key: IRDZCMUQNGEQEP-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)