missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thiazolyl)phenol, 97%

Catalog No. p-7030930
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3182703 1 g
AAH3182709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3182703 Supplier Thermo Scientific Chemicals Supplier No. H3182703

CAS: 81015-49-8 | C9H7NOS | 177.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81015-49-8
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD07368615
InChI Key LMMPCMRUGFBCPV-UHFFFAOYSA-N
Synonym 4-2-thiazolyl phenol, 4-1,3-thiazol-2-yl phenol, 4-thiazol-2-yl phenol, thiazole,2-4-trifluoromethyl phenyl, 4-3h-1,3-thiazol-2-ylidene cyclohexa-2,5-dien-1-one, 4-thiazol-2-yl-phenol, 4-thiazole-2-yl phenol, acmc-20a0c2, 4-thiazol-2-vl-phenol
PubChem CID 5354497
IUPAC Name 4-(3H-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=C2NC=CS2

Specifications

Melting Point 162°C to 165°C
Quantity 1 g
Formula Weight 177.28
Percent Purity 97%
Chemical Name or Material 4-(2-Thiazolyl)phenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only