missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridyl)benzaldehyde 95.0+%, TCI America™

Catalog No. P16711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16711G 1 g
P16715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16711G Supplier TCI America Supplier No. P16711G
Only null left

Chemical Identifiers

CAS 127406-56-8
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD01863537
InChI Key NMLYGLCBSFKJFI-UHFFFAOYSA-N
Synonym 4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
PubChem CID 1515240
IUPAC Name 4-(pyridin-2-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Melting Point 50°C
Color Yellow
Quantity 1 g
Formula Weight 183.21
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Pyridyl)benzaldehyde