missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridyl)benzaldehyde, 98%

Catalog No. p-7036986
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5549803 1 g
AAH5549806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5549803 Supplier Thermo Scientific Chemicals Supplier No. H5549803
Only null left

CAS: 127406-56-8 | C12H9NO | 183.21 g/mol

4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

Solubility
Soluble in Chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 127406-56-8
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD01863537
InChI Key NMLYGLCBSFKJFI-UHFFFAOYSA-N
Synonym 4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
PubChem CID 1515240
IUPAC Name 4-pyridin-2-ylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Melting Point 50°C to 52°C
Flash Point 113°C (235°F)
Quantity 1 g
Solubility Information Soluble in Chloroform
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material 4-(2-Pyridyl)benzaldehyde
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only