missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Propyn-1-yl)morpholine 98.0+%, TCI America™

Supplier:  TCI America P21701G

 View more versions of this product

Catalog No. P21701G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(2-Propyn-1-yl)morpholine
Yellow
C7H11NO
1 g
4-Propargylmorpholine
C#CCN1CCOCC1
125.171
125.17
Liquid
5799-76-8
186°C
MFCD00510919
1993
OKDZHAQIKCVKFE-UHFFFAOYSA-N
4-prop-2-ynylmorpholine
412941
≥98.0% (GC)

Chemical Identifiers

5799-76-8
125.171
OKDZHAQIKCVKFE-UHFFFAOYSA-N
412941
C#CCN1CCOCC1
C7H11NO
MFCD00510919
4-Propargylmorpholine
4-prop-2-ynylmorpholine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions